Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glut4 Rabbit anti-Human, Mouse, Rat, Plant, PE-Cyanine7, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP149533PECY7
Description
Glut4 Polyclonal antibody specifically detects Glut4 in Human, Mouse, Rat, Plant samples. It is validated for Flow Cytometry, KnockDown.Specifications
Glut4 | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
GLUT-4, GLUT4Glucose transporter type 4, insulin-responsive, insulin-responsive glucose transporter type 4, solute carrier family 2 (facilitated glucose transporter), member 4, solute carrier family 2, facilitated glucose transporter member 4 | |
Rabbit | |
IgG | |
0.1 mL | |
Store at 4°C in the dark. Do not freeze. | |
Polyclonal | |
Human, Mouse, Plant, Rat |
Flow Cytometry, KnockDown | |
Flow Cytometry, KnockDown Validated | |
Affinity Purified, Conjugated | |
SLC2A4 | |
A synthetic peptide from the human Glucose Transporter GLUT4 protein [UniProt P14672] | |
Immunogen affinity purified | |
Cellular Markers, Core ESC Like Genes, Diabetes Research, Hematopoietic Stem Cell Markers, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neuroendocrinology, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers | |
Primary | |
6517.0 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title