Learn More
GLUT4 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 210481AP
Catalog No. 50-173-3227
Add to cart
Description
Description
This gene is a member of the solute carrier family 2 (facilitated glucose transporter) family and encodes a protein that functions as an insulin-regulated facilitative glucose transporter. In the absence of insulin, this integral membrane protein is sequestered within the cells of muscle and adipose tissue. Within minutes of insulin stimulation, the protein moves to the cell surface and begins to transport glucose across the cell membrane. Mutations in this gene have been associated with noninsulin-dependent diabetes mellitus (NIDDM).
Specifications
Specifications
GLUT4
0.19 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P14672
GLUT 4, GLUT4, SLC2A4
Rabbit
IgG
150 μL
-20°C
Primary
6517
Immunohistochemistry (Paraffin)
Unconjugated
SLC2A4
Liquid
SLC2A4
GLUT4 Fusion Protein Ag15390
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human