Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glut5 Mouse anti-Human, Monkey, Janelia Fluor™ 549, Clone: OTI9F3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271285JF549

Catalog No. N271285J549


Add to cart

Description

Description

Glut5 Monoclonal antibody specifically detects Glut5 in Human, Monkey samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications

Specifications

Glut5
OTI9F3
Flow Cytometry, Immunohistochemistry
Affinity Purified, Conjugated
SLC2A5
Full length recombinant protein of human Glut5 (NP_003030) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human, Monkey
Flow Cytometry, Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
Fructose transporter, Glucose transporter type 5, small intestine, GLUT-5, GLUT5glucose transporter-like protein 5, solute carrier family 2 (facilitated glucose transporter), member 5, solute carrier family 2 (facilitated glucose/fructose transporter), member 5, solute carrier family 2, facilitated glucose transporter member 5
Mouse
IgG1
0.1 mL
Primary
6518.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit