Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutaconic aldehyde dianilide hydrochloride, mixture of isomers, 98%, Thermo Scientific Chemicals

Catalog Number p-7024147
Click to view available options
Quantity:
5 g
25 g
100 g
1497-49-0
C17H17ClN2
284.79
MFCD00012630
VUCMMJBDNXZQDJ-ZUJIUJENSA-N
hydrogen n-1e,3e-5-phenylimino penta-1,3-dien-1-yl aniline chloride
5702667
N-[(1E,3E)-5-phenyliminopenta-1,3-dienyl]aniline;hydrochloride
[H+].[Cl-].N(\C=C\C=C\C=NC1=CC=CC=C1)C1=CC=CC=C1
1497-49-0
C17H17ClN2
284.79
MFCD00012630
VUCMMJBDNXZQDJ-ZUJIUJENSA-N
hydrogen n-1e,3e-5-phenylimino penta-1,3-dien-1-yl aniline chloride
5702667
N-[(1E,3E)-5-phenyliminopenta-1,3-dienyl]aniline;hydrochloride
[H+].[Cl-].N(\C=C\C=C\C=NC1=CC=CC=C1)C1=CC=CC=C1

Glutaconic aldehyde dianilide hydrochloride is a tricarbocyanine laser dye intermediate. It is used as an intermediate for pharmaceuticals and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glutaconic aldehyde dianilide hydrochloride is a tricarbocyanine laser dye intermediate. It is used as an intermediate for pharmaceuticals and dyes.

Solubility
Soluble in methanol. Insoluble in water..

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature.
TRUSTED_SUSTAINABILITY
Melting Point ∼124°C to 132°C
Quantity 5 g
Beilstein 3916924
Sensitivity Hygroscopic
Solubility Information Soluble in methanol. Insoluble in water..
Formula Weight 284.79
Percent Purity 98%
Chemical Name or Material Glutaconic aldehyde dianilide hydrochloride, mixture of isomers
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 216-094-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit