Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutamate Receptor 6 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP30366720UL

 View more versions of this product

Catalog No. NB093667


Add to cart

Description

Description

Glutamate Receptor 6 Polyclonal antibody specifically detects Glutamate Receptor 6 in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence
Specifications

Specifications

Glutamate Receptor 6
Unconjugated
Purified
Rabbit
IgG
20 μL
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse
Immunofluorescence, Western Blot
PBS with 50% glycerol, pH7.3.
EAA4, glutamate receptor 6, glutamate receptor, ionotropic, kainate 2, ionotropic kainate 2, MGC74427
Recombinant fusion protein containing a sequence corresponding to amino acids 30-300 of human Glutamate Receptor 6 (NP_068775.1).
Affinity purified
RUO
Primary
2898
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit