Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutamate Receptor 6 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen Glutamate Receptor 6
Applications Immunofluorescence, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093666
View Documents
Novus Biologicals
NBP303667100UL
100 μg Each for $320.71
Add to cart
 
NB093667
View Documents
Novus Biologicals
NBP30366720UL
20 μg Each for $138.37
Add to cart
 
Description

Description

Glutamate Receptor 6 Polyclonal antibody specifically detects Glutamate Receptor 6 in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence
Specifications

Specifications

Glutamate Receptor 6
Unconjugated
Rabbit
Human, Mouse
EAA4, glutamate receptor 6, glutamate receptor, ionotropic, kainate 2, ionotropic kainate 2, MGC74427
Recombinant fusion protein containing a sequence corresponding to amino acids 30-300 of human Glutamate Receptor 6 (NP_068775.1).
Polyclonal
Affinity purified
Immunofluorescence, Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
2898
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit