Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutaminase Mouse anti-Human, Mouse, DyLight 405, Clone: 3211, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243762V

Catalog No. NBP243762V


Add to cart

Description

Description

Glutaminase Monoclonal antibody specifically detects Glutaminase in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Glutaminase
3211
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Purified
GLS
The exact sequence is proprietary.
Protein G purified
Store at 4C in the dark.
Monoclonal
Specific for GAC but not KGA isoform.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
AAD20, EC 3.5.1.2, GLS1DKFZp686O15119, glutaminase, glutaminase C, glutaminase kidney isoform, mitochondrial, glutaminase, phosphate-activated, K-glutaminase, KIAA0838FLJ10358, L-glutamine amidohydrolase
Mouse
IgG1
0.1 ml
Primary
2744
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit