Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Glutamine Synthetase Human Recombinant Protein, TMW: 42kDa
Highly purified and high bioactivity. Generating reliable and reproducible results.
$206.00 - $436.00
Specifications
Concentration | 1mg/mL |
---|---|
For Use With (Application) | SDS-Page |
Formulation | 20mM Tris-HCl buffer (pH8.0) containing 10% glycerol, 1mM DTT, 0.1mM PMSF |
Gene ID (Entrez) | 2752 |
Molecular Weight | TMW: 42kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2523762
|
Novus Biologicals™
NBP2523760.02MG |
0.02mg |
Each for $206.00
|
|
NBP25237601
|
Novus Biologicals™
NBP2523760.1MG |
0.1mg |
Each for $436.00
|
|
Description
An un-tagged recombinant protein corresponding to the amino acids 1-373 of Human Glutamine Synthetase The Recombinant Human Glutamine Synthetase Protein is derived from E. coli. The Recombinant Human Glutamine Synthetase Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.Specifications
1mg/mL | |
20mM Tris-HCl buffer (pH8.0) containing 10% glycerol, 1mM DTT, 0.1mM PMSF | |
TMW: 42kDa | |
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. | |
Glutamine Synthetase | |
Unlabeled |
SDS-Page | |
2752 | |
>85%, by SDS-PAGE | |
cell proliferation-inducing protein 59, EC 4.1.1.15, EC 6.3.1.2, GLNSPIG59, Glutamate decarboxylase, glutamate-ammonia ligase, Glutamate--ammonia ligase, glutamate-ammonia ligase (glutamine synthase), glutamine synthase, glutamine synthetase, GS, PIG43, proliferation-inducing protein 43 | |
Specific activity is > 2.000 pmol/min/ug, and is defined as the amount of enzyme that convert L-glutamate to L-glutamine per miunte at pH 7.5 at 37C coupled system with PK/LDH. | |
Yes |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title