Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutamine Synthetase Mouse anti-Human, PE/Cy5.5, Clone: OTI1F4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270834PECY55

Catalog No. N270834PEC5


Add to cart

Description

Description

Glutamine Synthetase Monoclonal antibody specifically detects Glutamine Synthetase in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Glutamine Synthetase
OTI1F4
Flow Cytometry
Affinity Purified, Conjugated
GLUL
Full length recombinant protein of human Glutamine Synthetase (NP_002056) produced in HEK293T cells.
Immunogen affinity purified
Hypoxia, Neurodegeneration, Neuroscience, Vision
Primary
2752.0
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
cell proliferation-inducing protein 59, EC 4.1.1.15, EC 6.3.1.2, GLNSPIG59, Glutamate decarboxylase, glutamate-ammonia ligase, Glutamate--ammonia ligase, glutamate-ammonia ligase (glutamine synthase), glutamine synthase, glutamine synthetase, GS, PIG43, proliferation-inducing protein 43
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit