Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutaraldehyde, 25% Aqueous Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
19L
Packaging:
Narrow Mouth Amber Glass Bottle
Poly Pail

Chemical Identifiers

111-30-8
C5H8O2
100.117
SXRSQZLOMIGNAQ-UHFFFAOYSA-N
3485
CHEBI:64276
pentanedial
C(CC=O)CC=O

High quality chemicals for laboratory and specialized industrial use

Quantity 19L
Formula Weight 100.12
Packaging Poly Pail
Specific Gravity 1L = 1.09kg
Chemical Name or Material Glutaraldehyde
Assay 25% Aqueous Solution
Grade BAKER ANALYZED™ Reagent