Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutaraldehyde Solution, 50% (Certified), Fisher Chemical

Click to view available options
Quantity:
1L
Packaging:
Glass Bottle
111-30-8, 7732-18-5
C5H8O2
100.12
MFCD00007025
SXRSQZLOMIGNAQ-UHFFFAOYSA-N
glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
3485
CHEBI:64276
pentanedial
O=CCCCC=O
111-30-8, 7732-18-5
C5H8O2
100.12
MFCD00007025
SXRSQZLOMIGNAQ-UHFFFAOYSA-N
glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
3485
CHEBI:64276
pentanedial
O=CCCCC=O
Quantity 1L
Additional Information Vapor Pressure: 15mmHg at 20°C
Clarity Pass Test
Density 1.130g/cm³
Identification Pass Test
Assay Percent Range 49.0 to 52.0 %
Packaging Glass Bottle
Linear Formula HC(O)(CH2)3CHO
UN Number 2927
Specific Gravity 1.125 to 1.135 (at 20°C)
Vapor Pressure 15mmHg at 20°C
Viscosity 20 mPa/s (50°C)
Formula Weight 100.12
Physical Form Liquid
Decomposition Information Decomposition Information: 110°C
Percent Purity 49.0 to 52.0%
Grade Certified
Boiling Point 101.5°C
Melting Point -33°C
pH 3.7
Chemical Name or Material Glutaraldehyde Solution, 50%
Show More Show Less

DANGER!

Emergency Overview
Toxic by inhalation and if swallowed. Causes burns by all exposure routes. May cause allergic respiratory and skin reaction. Very toxic to aquatic organisms.
NFPA
Health: 3
Flammability: 1
Instability: 1

SIGNAL WORD: DANGER!

Recommended Storage : Refrigerate