Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutaric anhydride, 90%, ACROS Organics™

Click to view available options
Quantity:
50g
250g
1kg
Packaging:
Glass bottle
108-55-4
C5H6O3
114.1
MFCD00006679
VANNPISTIUFMLH-UHFFFAOYSA-N
glutaric anhydride,dihydro-2h-pyran-2,6 3h-dione,glutaric acid anhydride,pentanedioic anhydride,pyroglutaric acid,2h-pyran-2,6 3h-dione, dihydro,pentanedioic acid anhydride,anhydrid kyseliny glutarove,unii-63ofi15s80,anhydrid kyseliny glutarove czech
7940
oxane-2,6-dione
C1CC(=O)OC(=O)C1
108-55-4
C5H6O3
114.1
MFCD00006679
VANNPISTIUFMLH-UHFFFAOYSA-N
glutaric anhydride,dihydro-2h-pyran-2,6 3h-dione,glutaric acid anhydride,pentanedioic anhydride,pyroglutaric acid,2h-pyran-2,6 3h-dione, dihydro,pentanedioic acid anhydride,anhydrid kyseliny glutarove,unii-63ofi15s80,anhydrid kyseliny glutarove czech
7940
oxane-2,6-dione
C1CC(=O)OC(=O)C1
Packaging Glass bottle
Quantity 1kg
Color Beige to Brown
Infrared Spectrum Authentic
Melting Point 46°C to 57°C
Assay Percent Range 90%
Formula Weight 114.1
Physical Form Crystalline Powder
Percent Purity ≥88% (morpholine coupling)
Chemical Name or Material Glutaric anhydride

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning