Learn More
Glutaric anhydride, 90%, ACROS Organics™
Specifications
Packaging | Glass Bottle |
Infrared Spectrum | Authentic |
Color | Beige to Brown |
Melting Point | 46°C to 57°C |
Quantity | 250g |
Assay Percent Range | 90% |
Formula Weight | 114.1 |
Physical Form | Crystalline Powder |
Percent Purity | ≥88% (morpholine coupling) |
Chemical Name or Material | Glutaric anhydride |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning