Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutaric anhydride, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass bottle
Plastic bottle
108-55-4
C5H6O3
114.1
MFCD00006679
VANNPISTIUFMLH-UHFFFAOYSA-N
glutaric anhydride,dihydro-2h-pyran-2,6 3h-dione,glutaric acid anhydride,pentanedioic anhydride,pyroglutaric acid,2h-pyran-2,6 3h-dione, dihydro,pentanedioic acid anhydride,anhydrid kyseliny glutarove,unii-63ofi15s80,anhydrid kyseliny glutarove czech
7940
oxane-2,6-dione
C1CC(=O)OC(=O)C1
108-55-4
C5H6O3
114.1
MFCD00006679
VANNPISTIUFMLH-UHFFFAOYSA-N
glutaric anhydride,dihydro-2h-pyran-2,6 3h-dione,glutaric acid anhydride,pentanedioic anhydride,pyroglutaric acid,2h-pyran-2,6 3h-dione, dihydro,pentanedioic acid anhydride,anhydrid kyseliny glutarove,unii-63ofi15s80,anhydrid kyseliny glutarove czech
7940
oxane-2,6-dione
C1CC(=O)OC(=O)C1
Melting Point 46.0°C to 57.0°C
Quantity 5g
Packaging Glass bottle
Boiling Point 290.0°C
Color White
Infrared Spectrum Authentic
Flash Point 173°C
Assay Percent Range 94% min. (GC)
Beilstein 17, V,11, 9
Solubility Information Solubility in water: hydrolysis. Other solubilities: soluble in benzene, toluene, acetone and, chloroform
Formula Weight 114.1
Physical Form Crystalline Powder or Crystals
Percent Purity 95%
Chemical Name or Material Glutaric anhydride, 97%
Show More Show Less

GHS H Statement
Causes serious eye damage.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:

GHS Signal Word: Danger

EINECSNumber : 203-593-6

RTECSNumber : MA3850000

TSCA : TSCA