Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutarimide, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
1121-89-7
C5H7NO2
113.116
MFCD00006670
KNCYXPMJDCCGSJ-UHFFFAOYSA-N
glutarimide,2,6-piperidinedione,2,6-diketopiperidine,2,6-dioxopiperidine,acetylglycinamide impurity b,piperidin-2,6-dione,acmc-2099da,5-21-09-00557 beilstein handbook reference,2,6-piperidinedione 9ci
70726
CHEBI:5435
piperidine-2,6-dione
C1CC(=O)NC(=O)C1
1121-89-7
C5H7NO2
113.116
MFCD00006670
KNCYXPMJDCCGSJ-UHFFFAOYSA-N
glutarimide,2,6-piperidinedione,2,6-diketopiperidine,2,6-dioxopiperidine,acetylglycinamide impurity b,piperidin-2,6-dione,acmc-2099da,5-21-09-00557 beilstein handbook reference,2,6-piperidinedione 9ci
70726
CHEBI:5435
piperidine-2,6-dione
C1CC(=O)NC(=O)C1
Packaging Glass bottle
Melting Point 154°C to 157°C
Quantity 100g
Infrared Spectrum Authentic
Color White
Assay Percent Range 98%
Beilstein 21, 382
Formula Weight 113.12
Physical Form Crystals or Crystalline Flakes
Percent Purity ≥97.5%
Chemical Name or Material Glutarimide, 98%
Show More Show Less

EINECSNumber : 214-340-4

RTECSNumber : MA4000000