Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Glutaronitrile, 99%

Click to view available options
Quantity:
50g
544-13-8
C5H6N2
94.117
MFCD00001970
ZTOMUSMDRMJOTH-UHFFFAOYSA-N
glutaronitrile,1,3-dicyanopropane,glutarodinitrile,glutaric acid dinitrile,1,3-trimethylenedinitrile,pyrotartaric acid nitrile,glutaronitrle,unii-01zi68f3cq,trimethylene dicyanide,glutaric dinitrile
10994
pentanedinitrile
C(CC#N)CC#N
544-13-8
C5H6N2
94.117
MFCD00001970
ZTOMUSMDRMJOTH-UHFFFAOYSA-N
glutaronitrile,1,3-dicyanopropane,glutarodinitrile,glutaric acid dinitrile,1,3-trimethylenedinitrile,pyrotartaric acid nitrile,glutaronitrle,unii-01zi68f3cq,trimethylene dicyanide,glutaric dinitrile
10994
pentanedinitrile
C(CC#N)CC#N
Applications
It can be used for the conversion, by exchange, of carboxylic acids to nitriles, by heating in the presence of phosphoric acid. The equilibrium is displaced by cyclization to glutarimide. It is also used in the preparation of dicarbonyl compounds.

Solubility
Soluble in water, alcohol and chloroform.

Notes
Hygroscopic. Protect from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Density 0.99
Refractive Index 1.435
Quantity 50g
Flash Point 112°C (234°F)
Boiling Point 286°C to 288°C
Melting Point -29°C
UN Number UN3276
Beilstein 1738385
Merck Index 14,4474
Solubility Information Soluble in water,alcohol and chloroform.
Formula Weight 94.12
Percent Purity 99%
Chemical Name or Material Glutaronitrile
Show More Show Less

GHS H Statement
H302
Harmful if swallowed.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H312+H332-H315-H319

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

EINECSNumber : 208-861-6

RTECSNumber : YI3500000

TSCA : Yes

Recommended Storage : Ambient temperatures