Alfa Aesar™ Glutaronitrile, 99%

Manufacturer: Alfa Aesar™ B2366309

Quantity 10g

 View more versions of this product

Catalog No. AAB2366309

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 544-13-8
    Molecular Formula C5H6N2
    Formula Weight 94.12
    MDL Number MFCD00001970
    UN Number UN3276
    Beilstein 1738385
    Merck Index 14,4474
    Synonym 1,3-Dicyanopropane; Pentanedinitrile
    Chemical Name or Material Glutaronitrile

    Specifications

    Formula Weight 94.12
    Melting Point −29°C
    Flash Point 112°C (234°F)
    TSCA TSCA
    MDL Number MFCD00001970
    UN Number UN3276
    Merck Index 14,4474
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 286° to 288°C
    RTECS Number YI3500000
    Molecular Formula C5H6N2
    CAS 544-13-8
    Beilstein 1738385
    Synonym 1,3-Dicyanopropane; Pentanedinitrile
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
    Quantity 10g
    Chemical Name or Material Glutaronitrile
    Assay 99%

    Description

    Applications
    It can be used for the conversion, by exchange, of carboxylic acids to nitriles, by heating in the presence of phosphoric acid. The equilibrium is displaced by cyclization to glutarimide. It is also used in the preparation of dicarbonyl compounds.

    Solubility
    Soluble in water, alcohol and chloroform.

    Notes
    Hygroscopic. Protect from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit