Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutaryl-L-carnitine-(N-methyl-d3) lithium salt, ≥95% (TLC), MilliporeSigma™ Supelco™

$113.30 - $451.00

Specifications

CAS 1803252-72-3 (free acid)
Percent Purity ≥95% (TLC)
Grade Analytical Standard
Linear Formula C12D3H18 NO6 · xLi+
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-100-2284
View Documents
MilliporeSigma Supelco
953341MG
1 mg
Each for $113.30
Add to cart
 
11-100-2283
View Documents
MilliporeSigma Supelco
953345MG
5 mg
Each for $226.00
Add to cart
 
11-100-2282
View Documents
MilliporeSigma Supelco
9533410MG
10 mg
Each for $451.00
Add to cart
 
Description

Description

Specifications

Specifications

1803252-72-3 (free acid)
≥95% (TLC)
Analytical Standard
C12D3H18 NO6 · xLi+
Safety and Handling

Safety and Handling

ShelfLife : Limited shelf life, expiry date on the label

Recommended Storage : -20°C

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit