Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutathione Peroxidase 4/GPX4 Antibody (JU11-31), Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

Supplier:  Novus Biologicals NBP275511

Catalog No. NBP275511


Only null left
Add to Cart

Description

Description

Glutathione Peroxidase 4/GPX4 Monoclonal specifically detects Glutathione Peroxidase 4/GPX4 in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Glutathione Peroxidase 4/GPX4
Monoclonal
1 mg/mL
Western Blot 1:500-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:500
EC 1.11.1, EC 1.11.1.12, Glutathione peroxidase 4, glutathione peroxidase 4 (phospholipid hydroperoxidase), GPx-4, GSHPx-4, MCSP, PHGPxsnGPx, phospholipid hydroperoxidase, phospholipid hydroperoxide glutathione peroxidase, mitochondrial, snPHGPx, sperm nucleus glutathione peroxidase
Rabbit
22 kDa
100 μL
2879.0
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
JU11-31
Unconjugated
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
GPX4
Synthetic peptide within Human Glutathione Peroxidase 4/GPX4 aa 23-72/ 197. (SwissProt: P36969 Human; SwissProt: O70325 Mouse; SwissProt: P36970 Rat)
Protein A purified
Primary
Human, Mouse, Rat, Zebrafish
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.