Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutathione S-Transferase mu 1/GSTM1 Mouse anti-Human, Clone: CPTC-GSTMu1-3, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Glutathione S-Transferase mu 1/GSTM1
Clone CPTC-GSTMu1-3
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Applications Western Blot, Immunohistochemistry (Paraffin), Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122163
View Documents
Novus Biologicals
NBP30734510UG
100 μg
Each for $352.89
Add to cart
 
NB122164
View Documents
Novus Biologicals
NBP30734520UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

Glutathione S-Transferase mu 1/GSTM1 Monoclonal antibody specifically detects Glutathione S-Transferase mu 1/GSTM1 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin), Protein Array
Specifications

Specifications

Glutathione S-Transferase mu 1/GSTM1
0.2 mg/ml
Western Blot, Immunohistochemistry (Paraffin), Peptide Array
Purified
RUO
Human
EC 2.5.1.18, glutathione S-alkyltransferase, glutathione S-aralkyltransferase, glutathione S-aryltransferase, glutathione S-transferase M1, glutathione S-transferase mu 1, GST class-mu 1, GST HB subunit 4, GST1, GSTM1-1, GSTM1a-1a, GSTM1b-1b, GTH4, GTM1, H-B, HB subunit 4, MGC26563, MU, MU-1, S-(hydroxyalkyl)glutathione lyase
Recombinant human full-length Glutathione S-Transferase mu 1/GSTM1 protein
Monoclonal
Protein A or G purified
CPTC-GSTMu1-3
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Unconjugated
Mouse
Asthma, Cancer, Immunology
10 mM PBS with 0.05% BSA
2944
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit