Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutathione S-Transferase pi 1/GSTP1 Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI4B6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270857AF647

Catalog No. N270857A647


Add to cart

Description

Description

Glutathione S-Transferase pi 1/GSTP1 Monoclonal antibody specifically detects Glutathione S-Transferase pi 1/GSTP1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Glutathione S-Transferase pi 1/GSTP1
OTI4B6
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
GSTP1
Full length recombinant protein of human Glutathione S-Transferase pi 1/GSTP1 (NP_000843) produced in E.coli.
Immunogen affinity purified
Asthma, Cancer, Immunology, Neuroscience
Primary
2950.0
Immunohistochemistry, Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
deafness, X-linked 7, EC 2.5.1.18, FAEES3, fatty acid ethyl ester synthase III, glutathione S-transferase P, glutathione S-transferase pi 1, GST class-pi, GST3DFN7, GSTP, GSTP1-1, PI
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit