Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Glutathione Sepharose™ 4 FF Media PROMO

Single-step purification of GST-tagged proteins and other glutathione S-transferase or glutathione-dependent proteins. Glutathione Sepharose 4 Fast Flow Media is used for the purification of GST-tagged proteins from bacterial lysates and other S-tranferases or glutathione-dependent proteins.

Manufacturer:  GE Healthcare 29361661

Catalog No. 45-000-286PM

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

  • Glutathione coupled to Sepharose 4 Fast Flow via 10-carbon spacer arm
  • Protein binding capacity: »10mg recombinant GST/mL media (flowrate dependent)
  • Binding capacity and high flow properties allow scale-up
  • Mild elution conditions preserve protein antigenicity and function
  • Stable in all commonly used aqueous buffers
  • pH stability: 3 to 12

Specifications

Specifications

>10 mg/mL
<450cm/hr.
Glutathione and 10-carbon linker arm
100 mL
90μm
base matrix 150-250 cm/h, 100 kPa, XK 50/60 column, bed height 25 cm
4 to 30°C, 20% Ethanol
Glutathione Sepharose 4 Fast Flow is an affinity resin especially suited for purification of GST-tagged proteins in a batch mode or when scale up is needed.
10 Carbon spacer arm, 4% Agarose, 90μm particle size
Highly cross-linked 4% agarose
3 to 12
Glutathione Sepharose 4 Fast Flow, 100 mL
100mL
4% Highly Cross-linked Agarose
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit