Learn More
GE Healthcare Glutathione Sepharose™ 4B Media: PROMO
Use for high capacity purification of Glutathione S-Transferase (GST) tagged fusion proteins. GE Healthcare Life Sciences™ Glutathione Sepharose™ 4B Media requires only a single-step for purification.
Manufacturer:  GE Healthcare 17075601PM
Catalog No. 45-000-139LR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Binding capacity: >5mg horse liver GST/mL medium (flowrate dependent) Average particle size: 90μm
  • Recommended flowrate (H2O at room temp.): 75cm/hour
  • Stable in presence of all commonly used aqueous buffers
  • Stable in pH range 4 to 13
Specifications
Specifications
Glutathione Sepharose 4 B
90 µm
4 to 13
10 mL