Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Glutathione Sepharose™ 4B Media : PROMOTION

Promotion Details: Receive a 15% discount.

For single-step purification of recombinant derivatives of glutathione S-transferase, other glutathione S-transferases, and glutathione-dependent proteins

Manufacturer:  GE Healthcare 17075601PM/DEL

Catalog No. 45-000-139PM

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

  • Binding capacity: >5mg horse liver GST/mL medium (flowrate dependent) Average particle size: 90µm
  • Recommended flowrate (H2O at room temp.): 75cm/hour
  • Stable in presence of all commonly used aqueous buffers
  • Stable in pH range 4 to 13

Specifications

Specifications

Glutathione Sepharose 4 B
90 µm
4 to 13
10 mL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit