Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Glutathione Sepharose™ High Performance Media
For high resolution purification of Glutathione S transferase (GST) tagged proteins
$1222.00 - $3474.00
Specifications
Binding Capacity | >Approx. 10 mg GST-tagged protein/mL resin |
---|---|
Description | Glutathione Sepharose High Performance is an affinity resin recommended for purifying glutathione S-transferase (GST)-tagged proteins when high resolution is the priority. |
Flow Rate | <600 cm/hr. |
Matrix | Highly crossed-linked agarose, 6% |
pH Range | 3 to 12 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
45-000-331
|
Cytiva
17527902 |
100 mL |
Each for $3,474.00
|
|
|||||
45-000-330
|
Cytiva
17527901 |
25 mL |
Each for $1,222.00
|
|
|||||
Description
- One-step purification of GST-tagged proteins and other glutathione S-transferases or glutathione-dependent proteins
- GST-tagged proteins are eluted under mild, nondenaturing conditions using reduced glutathione
- Mild purification process preserves protein antigenicity and function
- Glutathione ligand is coupled to 6% agarose matrix
Specifications
>Approx. 10 mg GST-tagged protein/mL resin | |
<600 cm/hr. | |
3 to 12 | |
4°C to 30°C, 20% Ethanol | |
Chromatography Media |
Glutathione Sepharose High Performance is an affinity resin recommended for purifying glutathione S-transferase (GST)-tagged proteins when high resolution is the priority. | |
Highly crossed-linked agarose, 6% | |
6% Highly Cross-linked Agarose | |
34 μm | |
Glutathione |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title