Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutathione Synthetase Mouse anti-Human, Janelia Fluor™ 646, Clone: OTI1B8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270854JF646

Catalog No. N270854J646


Add to cart

Description

Description

Glutathione Synthetase Monoclonal antibody specifically detects Glutathione Synthetase in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications

Specifications

Glutathione Synthetase
OTI1B8
Flow Cytometry, Immunohistochemistry
Affinity Purified, Conjugated
GSS
Full length recombinant protein of human Glutathione Synthetase (NP_000169) produced in HEK293T cells.
Immunogen affinity purified
Neuroscience
Primary
2937.0
Flow Cytometry, Immunohistochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
EC 6.3.2.3, Glutathione synthase, glutathione synthetase, GSH synthetase, GSHS, GSH-S, MGC14098
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit