Learn More
Enzo Life SciencesSupplier Diversity Partner Glutathione (GSSG/GSH), detection kit (4x96wells)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADI900160
Rapid, ultra-sensitive kit for the quantification of reduced (GSH), oxidized (GSSG), and total glutathione for oxidative stress research. The Glutathione (GSSG/GSH), detection kit provides a highly sensitive enzymatic recycling reaction for the measurement of total, oxidized, or reduced glutathione in a convenient 96-well plate format. Sensitivity: 6.25 pmoles/well (range 12.5 - 100 pmoles/well). Applications: Colorimetric detection. Contents: Microtiter plate, Assay buffer concentrate, Reaction mix, Glutathione reductase, GSSG
Catalog No. 50-658-186