Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ UltraFIT™ Nylon Laboratory/Cleanroom Glove Liners

Catalog Number 1437936
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Pre-shrunk nylon glove liners protect extra-sensitive hands from irritation and occlusion of the skin associated with frequent gloving.

  • Pre-shrunk nylon is unaffected by repeated laundering
  • Seamless fingertips ensure optimum dexterity
  • Protects extra sensitive hands from irritation and occlusion of the skin
  • Use to provide an extra layer of warmth when working in cold areas
  • Color coded hems for easy size identification
Coating Material Uncoated
Color White
Disposable Reusable
Hand Ambidextrous
Material Nylon
Size Medium
For Use With (Application) Preventing Irritation and Skin Occlusion
Glove Cuff Style Knit
Series UltraFIT™
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit