Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gly-Gly, JustPURE™, white powder, Fisher BioReagents

Click to view available options
Quantity:
50g
556-50-3
Diglycine
556-50-3
Diglycine
Purity or Quality Grade JustPURE™
Color White
Format Powder/Solid
Molecular Weight 132.119
pH 5.5 to 6.0
Purity 99.5% min.
Quantity 50g
Packaging Poly Bottle
Solubility Soluble in water