Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLYAT Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548504

Catalog No. PA548504

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine, mouse and rat based on sequence homology.

Mitochondrial acyltransferase which transfers an acyl group to the N-terminus of glycine and glutamine, although much less efficiently. Can conjugate numerous substrates to form a variety of N-acylglycines, with a preference for benzoyl-CoA over phenylacetyl-CoA as acyl donors. Thereby detoxify xenobiotics, such as benzoic acid or salicylic acid, and endogenous organic acids, such as isovaleric acid.
Specifications

Specifications

GLYAT
Polyclonal
PBS with 0.09% sodium azide
Q6IB77
Glycine N-acyltransferase, Acyl-CoA:glycine N-acyltransferase, AAc, Aralkyl acyl-CoA N-acyltransferase, Aralkyl acyl-CoA:amino acid N-acyltransferase, Benzoyl-coenzyme A:glycine N-acyltransferase, Glycine N-benzoyltransferase, HRP-1(CLP), GLYAT, ACGNAT, CAT, GAT
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10249
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
GLYAT
Liquid
GLYAT
KLH conjugated synthetic peptide between 171-199 amino acids from the Central region of human GLYAT.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit