Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glyburide, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
10238-21-8
C23H28ClN3O5S
494.00
ZNNLBTZKUZBEKO-UHFFFAOYSA-N
5-chloro-N-[2-(4-{[(cyclohexylcarbamoyl)amino]sulfonyl}phenyl)ethyl]-2-methoxybenzamide
COC1=CC=C(Cl)C=C1C(=O)NCCC1=CC=C(C=C1)S(=O)(=O)NC(=O)NC1CCCCC1
10238-21-8
C23H28ClN3O5S
494.00
ZNNLBTZKUZBEKO-UHFFFAOYSA-N
5-chloro-N-[2-(4-{[(cyclohexylcarbamoyl)amino]sulfonyl}phenyl)ethyl]-2-methoxybenzamide
COC1=CC=C(Cl)C=C1C(=O)NCCC1=CC=C(C=C1)S(=O)(=O)NC(=O)NC1CCCCC1

G1130, 10238-21-8, C23H28ClN3O5S

Spectrum™ Glyburide, USP is an oral diabetes medicine that helps control blood sugar levels and is used to treat type 2 diabetes. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Ignition Residue 0.005
Quantity 5 g
Loss on Drying 0.01
CAS Max % 1
Percent Purity 98 to 102%