Learn More
Glycerides Model Set Products for Science Education
Explore the structure of glycerides with this kit.
Manufacturer:  Ryler Enterprises GLY1
Catalog No. S94028
Add to cart
Includes: Kit Contents:
  • 25 purple3-peg sugar
  • 25 yellow 2-peg phosphate
  • 9 red 3-peg adenine
  • 9 black 3-peg thymine
  • 5 green 4-peg cytosine
  • 5 silver 4-peg guanine
  • 75-1.25 in. PVC, clear bonds (single bonds)
  • 30-2 in. PVC, white bonds (H bonds)
  • Lab procedure
  • Answer key
Description
Description
  • Color-coded, modeled plastic atom centers are 1/4 in. diameter
  • Pegs are 3/8 in. long and are set at angles peculiar to the elements the atoms represent
  • Standard color-coding
  • Atom centers are bonded by connecting pegs with pvc tubing
Specifications
Specifications
Explore the structure of Glycerides
60 White 1-peg Hydrogen atom centers, 16 blue 4-peg Nitrogen atom centers, 5 red 4-peg Oxygen atom centers, 25 red 2-peg Oxygen atom centers, 40 black 4-peg Carbon atom centers, 3 purple 5-peg Phosphorous atom centers, One hundred twenty eight 1.25 in. PVC, clear bonds (single bonds), Thirty 1.625 in. PVC, clear bonds (double bonds), Six 2 in. PVC, white bonds (H bonds), Five 0.75 in. PVC, white bonds (Glycosidic bonds), lab procedure, answer key
Glycerides model set