Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerin, 100% Vegetable Based, U.S.P., J.T.Baker™

Click to view available options
Quantity:
500mL
4L
19L
Packaging:
Narrow Mouth Amber Glass Bottle
Poly Pail

Chemical Identifiers

56-81-5
C3H8O3
92.094
PEDCQBHIVMGVHV-UHFFFAOYSA-N
753
CHEBI:17754
propane-1,2,3-triol
C(C(CO)O)O
Quantity 19L
Formula Weight 92.1
Packaging Poly Pail
Specific Gravity 1L = 1.26kg
Flash Point 199.00°C
Chemical Name or Material Glycerin
Assay 100% Vegetable Based
Grade USP