Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerin, Multi-Compendial, U.S.P., J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle
56-81-5
C3H8O3
92.094
PEDCQBHIVMGVHV-UHFFFAOYSA-N
glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
753
CHEBI:17754
propane-1,2,3-triol
C(C(CO)O)O
56-81-5
C3H8O3
92.094
PEDCQBHIVMGVHV-UHFFFAOYSA-N
glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
753
CHEBI:17754
propane-1,2,3-triol
C(C(CO)O)O
Packaging Narrow Mouth Amber Glass Bottle
Flash Point 199.00°C
Specific Gravity 1L = 1.26kg
Quantity 4L
Formula Weight 92.1
Grade Multi-Compendial/USP
Chemical Name or Material Glycerin