Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerin, Natural, USP, 99-101%, Spectrum™

Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
Packaging:
Poly Bottle
Poly Drum
56-81-5
C3H8O3
92.09
PEDCQBHIVMGVHV-UHFFFAOYSA-N
propane-1,2,3-triol
OCC(O)CO
56-81-5
C3H8O3
92.09
PEDCQBHIVMGVHV-UHFFFAOYSA-N
propane-1,2,3-triol
OCC(O)CO

G1589, 56-81-5, C3H8O3

Spectrum™ Glycerin, Natural, USP is one of two USP grade Glycerins available from Spectrum with the other being Glycerin, Synthetic, USP, EP, BP, JP . Natural glycerin is produced from the hydrolysis of animal or vegetable lipids during soap manufacture but the majority of natural glycerin now comes as a result of it being a side product during the production of biodiesel. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Bottle
Ignition Residue 0.0001
Quantity 4 L
CAS Max % 1
Percent Purity 99 to 101%