Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerin, Sterile Reagent, BiotechGrade, Spectrum™

Click to view available options
Quantity:
100 mL
Packaging:
Poly Bottle

G3054, 56-81-5, C3H8O3

Spectrum™ Glycerin, Sterile Reagent, BiotechGrade , is a sugar alcohol (polyol) compound. An odorless, viscous and colorless liquid that has three hydroxyl groups that allow its solubility in water and give it it′s hygroscopic nature. Used in the personal care and pharmaceutical industry for improving smoothness and lubrication it is also used as a solvent and preservative. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.
Packaging Poly Bottle
Quantity 100 mL
Appearance Clear
CAS 56-81-5
CAS Max % 1
Molecular Formula C3H8O3
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
SMILES OCC(O)CO
IUPAC Name propane-1,2,3-triol
Molecular Weight (g/mol) 92.09