Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerin, Synthetic, EP, BP, JP, USP, 99-101%, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Poly Drum
56-81-5
C3H8O3
92.09
PEDCQBHIVMGVHV-UHFFFAOYSA-N
propane-1,2,3-triol
OCC(O)CO
56-81-5
C3H8O3
92.09
PEDCQBHIVMGVHV-UHFFFAOYSA-N
propane-1,2,3-triol
OCC(O)CO

G1015, 56-81-5, C3H8O3

Spectrum™ Glycerin, Synthetic, USP, EP, BP, JP is one of two USP grade Glycerins available from Spectrum with the other being Glycerin, Natural, USP, EP, BP. For those products requiring FDA approvals, if the initial formulation was done with Glycerin, Synthetic, USP, EP, BP, JP, the use of Glycerin, Natural, USP, EP, BP, JP is not interchangeable but would require the new formulation to undergo a new FDA approval process.
Packaging Poly Drum
Water 0.02
Ignition Residue 0.0001
Specific Gravity 1.258
Refractive Index 1.470-1.475
Quantity 20 L
Appearance Clear
CAS Max % 1
Percent Purity 99 to 101%