Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerin, Synthetic, FCC, 95-100.5%, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Poly Drum
56-81-5
C3H8O3
92.09
PEDCQBHIVMGVHV-UHFFFAOYSA-N
propane-1,2,3-triol
OCC(O)CO
56-81-5
C3H8O3
92.09
PEDCQBHIVMGVHV-UHFFFAOYSA-N
propane-1,2,3-triol
OCC(O)CO

G1040, 56-81-5, C3H8O3

Spectrum™ Glycerin, Synthetic, FCC is used in food and beverage, and personal care ingredients with the primary usages in sugar substitutes, moisturizing agent for baked goods and as a thickener in ear drops, toothpaste and jellies. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Drum
Ignition Residue 0.0001
Quantity 20 L
CAS Max % 1
Percent Purity 95 to 100.5%