Glycerine, Natural, GMP/ACS/USP/NF/FCC Grade, PHARMCO-AAPER

Manufacturer: Pharmco Products 349000GMPDM55M

Packaging Amber glass bottle
Quantity 55 gal.

 View more versions of this product

Catalog No. 16-101-432

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 56-81-5
    Physical Form Liquid
    Molecular Formula C3H8O3
    Formula Weight 92.09
    Synonym 1, 2, 3-Propanetriol
    Chemical Name or Material Gylcerine

    Specifications

    Formula Weight 92.09
    Quantity 55 gal.
    Color Clear
    Physical Form Liquid
    Molecular Formula C3H8O3
    CAS 56-81-5
    Synonym 1, 2, 3-Propanetriol
    Odor Odorless
    Packaging Amber glass bottle
    Stability Stable under recommended storage conditions.
    Chemical Name or Material Gylcerine

    Order Info

    Minimum Order Quantity: 5

    Minimum Order Quantity: 5

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit