Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Glycerol, 99.0 to 101.0%, Molecular Biology Grade, Ultrapure, Thermo Scientific™

Click to view available options
Quantity:
500mL
1L
Packaging:
Plastic bottle
56-81-5
C3H8O3
92.094
MFCD00004722
PEDCQBHIVMGVHV-UHFFFAOYSA-N
glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
753
CHEBI:17754
propane-1,2,3-triol
C(C(CO)O)O
56-81-5
C3H8O3
92.094
MFCD00004722
PEDCQBHIVMGVHV-UHFFFAOYSA-N
glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
753
CHEBI:17754
propane-1,2,3-triol
C(C(CO)O)O
  • Used for long-term storage of bacterial cultures
  • Step gradients for bacteriophage/particle purification
  • Mammalian cell transfection
  • Storage buffers for enzymes
Melting Point 18°C
Specific Gravity 1.26
Flash Point 160°C (320°F)
Packaging Plastic bottle
Boiling Point 182°C (20mmHg)
Quantity 1L
Linear Formula HOCH2CH(OH)CH2OH
Formula Weight 92.09
Physical Form Viscous Liquid
Percent Purity 99.0 to 101.0%
Grade Molecular Biology, Ultrapure
Chemical Name or Material Glycerol
Show More Show Less