Glycerol, ACS, 99.5%, Alfa Aesar

Manufacturer: Alfa Aesar 36646M1

Quantity 4 x 1L

 View more versions of this product

Catalog No. AA36646M1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 56-81-5
    Physical Form Viscous Liquid
    Assay Percent Range >99.5%
    Molecular Formula C3H8O3
    Formula Weight 92.09
    MDL Number MFCD00004722
    Chemical Name or Material Glycerol
    Grade ACS Grade

    Specifications

    Physical Form Viscous Liquid
    Formula Weight 92.09
    Quantity 4 x 1L
    EINECS Number 200-289-5
    Appearance Viscous liquid
    Assay Percent Range >99.5%
    Molecular Formula C3H8O3
    MDL Number MFCD00004722
    CAS 56-81-5
    Ignition Point 400°C (752°F)
    Health Hazard 1 H319: Causes serious eye irritation.
    Vapor Pressure <0.1 hPa (0mm Hg) at 20°C
    Chemical Name or Material Glycerol
    Grade ACS Grade

    H319: Causes serious eye irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit