Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerol, Anhydrous, Ultrapure Bioreagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
HDPE Pressure Bottom Poly Bottle
Poly Bottle
56-81-5
C3H8O3
92.094
PEDCQBHIVMGVHV-UHFFFAOYSA-N
glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
753
CHEBI:17754
propane-1,2,3-triol
C(C(CO)O)O
56-81-5
C3H8O3
92.094
PEDCQBHIVMGVHV-UHFFFAOYSA-N
glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
753
CHEBI:17754
propane-1,2,3-triol
C(C(CO)O)O

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For enzyme stabilization, freezing and electrophoresis buffers; For Laboratory,Research,or Manufacturing Use
Quantity 4L
Flash Point 199.00°C
Packaging Poly Bottle
Specific Gravity 1L = 1.26kg
Formula Weight 92.1
Grade Ultrapure Bioreagent
Chemical Name or Material Glycerol, Anhydrous