Glycerol, BAKER™, J.T.Baker™

A grade of chemicals for laboratory use

Manufacturer: J.T.Baker M77807

Packaging Narrow Mouth Amber Glass Bottle
Quantity 500mL

 View more versions of this product

Catalog No. 02-002-933

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 56-81-5
    Molecular Formula HOCH2CHOHCH2OH
    Formula Weight 92.1
    Chemical Name or Material Glycerol

    Specifications

    Formula Weight 92.1
    Flash Point 199.00°C
    CAS 56-81-5
    Packaging Narrow Mouth Amber Glass Bottle
    Specific Gravity 1L = 1.26kg
    Quantity 500mL
    Molecular Formula HOCH2CHOHCH2OH
    Chemical Name or Material Glycerol

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit