Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycerol Gel-Loading Dye, 5X (Contains 30% Glycerol/Molecular Biol.), Fisher BioReagents
$78.00 - $135.00

Specifications
Chemical Name or Material | Glycerol Gel-Loading Dye, 5X |
---|---|
Name Note | Contains 30% Glycerol |
CAS | 56-81-5, 7732-18-5 |
Grade | Molecular Biology |
Color | Colorless |
Catalog Number | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Quantity | Packaging | Price | Quantity & Availability | |||||
BP645-1
|
1mL | Poly Tube |
Each for $78.00
Order in Bulk
Request bulk or custom formats |
|
BP645-5
|
5mL | Poly Bottle |
Each for $135.00
Order in Bulk
Request bulk or custom formats |
|
Description
This dye is added to DNA and RNA samples prior to electrophoresis on agarose gels.Specifications
Glycerol Gel-Loading Dye, 5X | |
56-81-5, 7732-18-5 | |
Colorless |
Contains 30% Glycerol | |
Molecular Biology | |
Liquid |
Safety and Handling

Recommended Storage : RT
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title