Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerol (Glycerin), Purified, Reagents

Catalog No. p-200021167
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L

Glycerol (Glycerin), Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym 1,2,3-Propanetriol, Glycerine
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 18.2°C
Color Colorless
Boiling Point 544°C
Melting Range 18.2°C
Packaging Glass Bottle
Quantity 1 L
Specific Gravity 1.2613
Grade Pure
Physical Form Liquid
Chemical Name or Material Glycerol (Glycerin), Purified
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.