Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycerol Kinase Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP1780180.1MG
Description
Glycerol Kinase Polyclonal specifically detects Glycerol Kinase in Bacteria samples. It is validated for Western Blot, ELISA.Specifications
Glycerol Kinase | |
Polyclonal | |
Western Blot 1:500 - 1:2000, ELISA 1:4000 - 1:20000 | |
P04806 | |
GK | |
Glycerol Kinase [Cellulomonas species] | |
0.1 mg | |
Primary | |
Bacteria, Non-species specific | |
Purified |
Western Blot, ELISA | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
ATP:glycerol 3-phosphotransferase, EC 2.7.1.30, GK1, GKD, Glycerokinase, glycerol kinase | |
Goat | |
Ion exchange chromatography | |
RUO | |
2710 | |
Store at 4C. Do not freeze. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction