Learn More
MP Biomedicals™ Glycerol, Microbiological Grade
Glycerol, Microbiological Grade
Manufacturer:  MP Biomedicals™ 091004417
Catalog No. MP091004417
Add to cart
Description
Description
Highly purified for use as a fixative, a component in bacterial preservation media and for the isolation and maintenance of a variety of microorganisms.
Specifications
Specifications
Liquid
5.0-7.5
>99%
92.09
500g
Biochemicals
Room Temperature