Alfa Aesar™ Glycerol monostearate, purified

Manufacturer: Alfa Aesar™ 4388330

Quantity 250g

 View more versions of this product

Catalog No. AA4388330

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 31566-31-1
    Molecular Formula C17H35OCOCH2CH(OH)CH2OH
    Formula Weight 358.56
    MDL Number MFCD00036186
    Merck Index 14,4489
    Chemical Name or Material Glycerol monostearate, Purified

    Specifications

    Formula Weight 358.56
    TSCA TSCA
    Physical Form Powder
    Molecular Formula C17H35OCOCH2CH(OH)CH2OH
    MDL Number MFCD00036186
    CAS 31566-31-1
    Quantity 250g
    Merck Index 14,4489
    Solubility Information Soluble in hot organic solvents
    Chemical Name or Material Glycerol monostearate, Purified

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit