Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerol Solution, puriss., Meets analytical specification of Ph. Eur. BP, 84 to 88%, Honeywell™ Riedel-de Haen™

Click to view available options
Quantity:
1L
5L
25L
4 x 5L
6 x 1L
70kg
Packaging:
Poly Bottle
Poly Pail
Stainless Steel Can
56-81-5
C3H8O3
92.094
MFCD00004722
PEDCQBHIVMGVHV-UHFFFAOYSA-N
glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
753
CHEBI:17754
propane-1,2,3-triol
C(C(CO)O)O
56-81-5
C3H8O3
92.094
MFCD00004722
PEDCQBHIVMGVHV-UHFFFAOYSA-N
glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
753
CHEBI:17754
propane-1,2,3-triol
C(C(CO)O)O

Puriss., meets analytical specification of BP, European Pharmacopoeia, 84-88%

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
Quantity 70kg
Color Colorless
Refractive Index n20/D 1.449-1.455
Ignition Residue ≤0.01% (as SO4)
Packaging Poly Pail
Density 1.252g/mL at 25°C
Boiling Point 290°C
Flash Point 160°C (320°F)
Linear Formula HOCH2CH(OH)CH2OH · aq
UN Number NONH for all modes of transport
Beilstein 635685
Formula Weight 92.09g/mol (anhydrous basis)
Physical Form Liquid
Percent Purity 84 to 88%
Grade puriss.
Chemical Name or Material Glycerol Solution, Meets analytical specification of Ph. Eur. BP
Show More Show Less

EINECSNumber : 200-289-5

RTECSNumber : MA8050000