Alfa Aesar™ Glycerol tributyrate, 98%

Manufacturer: Alfa Aesar™ A1183030

Quantity 250g

 View more versions of this product

Catalog No. AAA1183030

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 60-01-5
    Molecular Formula C15H26O6
    Formula Weight 302.37
    MDL Number MFCD00009392
    Beilstein 1714746
    Merck Index 14,9620
    Synonym Tributyrin
    Chemical Name or Material Glycerol tributyrate

    Specifications

    Formula Weight 302.37
    CAS 60-01-5
    Melting Point −75°C
    Flash Point 174°C (345°F)
    TSCA TSCA
    Molecular Formula C15H26O6
    MDL Number MFCD00009392
    Merck Index 14,9620
    Synonym Tributyrin
    Boiling Point 307° to 308°C
    RTECS Number ET7350000
    Beilstein 1714746
    Quantity 250g
    Chemical Name or Material Glycerol tributyrate
    Assay 98%

    Description

    Applications
    Glycerol tributyrate is a triglyceride that may inhibit cell growth and induce cell differentiation. It is used to to induce differentiation of malignant cells followed by arrest of cell growth and apoptosis.

    Solubility
    Not miscible in water.

    Notes
    Incompatible with strong oxidizing agents, Strong acids, Strong bases. Use with adequate ventilation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit